siRNA Details
| Gene Symbol: Ren1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031192.3 | NP_112469.1 | 2 | 229 | ||
| GenBank mRNA | AF117821.1 | - | 2 | 171 | ||
| AK052685.1 | - | 2 | 229 | |||
| AK085309.1 | - | 2 | 229 | |||
| BC061053.1 | - | 2 | 227 | 30313659 | ||
| X16642.1 | - | 2 | 211 | |||

Ren1
Genomic Map