siRNA Details
| Gene Symbol: Kdf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083916.1 | NP_001077385.1 | 2 | 355 | ||
| NM_133707.2 | NP_598468.2 | 2 | 296 | |||
| GenBank mRNA | AK007551.1 | - | 1 | 220 | ||
| AK158383.1 | - | 2 | 438 | |||
| BC006890.1 | - | 2 | 224 | 3599314 | ||
| BY781417.1 | - | 2 | 267 | |||

Kdf1
Genomic Map