siRNA Details
| Gene Symbol: Ano10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271873.1 | NP_001258802.1 | 2 | 227 | ||
| NM_133979.3 | NP_598740.1 | 2 | 227 | |||
| GenBank mRNA | AK043011.1 | - | 2 | 190 | ||
| AK046091.1 | - | 2 | 179 | |||
| AK079622.1 | - | 2 | 178 | |||
| AK080947.1 | - | 2 | 179 | |||
| AK138414.1 | - | 2 | 213 | |||
| BC099688.1 | - | 2 | 150 | 6313494 | ||

Ano10
Genomic Map