siRNA Details
| Gene Symbol: Ndufs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271443.1 | NP_001258372.1 | 5 | 297 | ||
| NM_001271444.1 | NP_001258373.1 | 5 | 537 | |||
| NM_144870.5 | NP_659119.2 | 5 | 540 | |||
| GenBank mRNA | AK135069.1 | - | 5 | 298 | ||
| AK149674.1 | - | 5 | 498 | |||
| AK153182.1 | - | 5 | 484 | |||
| AK153900.1 | - | 5 | 274 | |||
| AK164901.1 | - | 5 | 274 | |||
| AK167907.1 | - | 5 | 275 | |||
| BC021616.1 | - | 5 | 258 | 5132866 | ||
| BC086766.1 | - | 5 | 251 | 6820786 | ||

Ndufs8
Genomic Map