siRNA Details
| Gene Symbol: Ubtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145500.3 | NP_663475.1 | 2 | 289 | ||
| GenBank mRNA | AK078479.1 | - | 2 | 289 | ||
| BC016129.1 | - | 2 | 115 | 4920477 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubtd1| Gene Symbol: Ubtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145500.3 | NP_663475.1 | 2 | 289 | ||
| GenBank mRNA | AK078479.1 | - | 2 | 289 | ||
| BC016129.1 | - | 2 | 115 | 4920477 | ||