siRNA Details
| Gene Symbol: Olfr866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146558.2 | NP_666769.1 | 1 | 188 | ||
| GenBank mRNA | BC119369.1 | - | 1 | 188 | 8734118 | |
| BC137762.1 | - | 1 | 188 | 8860781 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr866| Gene Symbol: Olfr866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146558.2 | NP_666769.1 | 1 | 188 | ||
| GenBank mRNA | BC119369.1 | - | 1 | 188 | 8734118 | |
| BC137762.1 | - | 1 | 188 | 8860781 | ||