siRNA Details
Gene Symbol: Olfr583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146757.1 | NP_666968.1 | 1 | 504 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
