siRNA Details
| Gene Symbol: Olfr904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146801.2 | NP_667012.2 | 1 | 271 | ||
| GenBank mRNA | BC114635.1 | - | 1 | 270 | ||
| BC146614.1 | - | 1 | 271 | 40040862 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr904| Gene Symbol: Olfr904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146801.2 | NP_667012.2 | 1 | 271 | ||
| GenBank mRNA | BC114635.1 | - | 1 | 270 | ||
| BC146614.1 | - | 1 | 271 | 40040862 | ||