siRNA Details
Gene Symbol: Olfr895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146875.2 | NP_667086.2 | 1 | 135 | ||
GenBank mRNA | BC127996.1 | - | 1 | 139 | ||
BC152319.1 | - | 1 | 139 | 40135771 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146875.2 | NP_667086.2 | 1 | 135 | ||
GenBank mRNA | BC127996.1 | - | 1 | 139 | ||
BC152319.1 | - | 1 | 139 | 40135771 |