siRNA Details
| Gene Symbol: Olfr1299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146884.2 | NP_667095.2 | 2 | 291 | ||
| GenBank mRNA | CB173344.1 | - | 2 | 291 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1299| Gene Symbol: Olfr1299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146884.2 | NP_667095.2 | 2 | 291 | ||
| GenBank mRNA | CB173344.1 | - | 2 | 291 | ||