siRNA Details
Gene Symbol: Olfr347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146943.1 | NP_667154.1 | 1 | 519 | ||
GenBank mRNA | BC127977.1 | - | 1 | 523 | 40135187 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146943.1 | NP_667154.1 | 1 | 519 | ||
GenBank mRNA | BC127977.1 | - | 1 | 523 | 40135187 |