siRNA Details
| Gene Symbol: Olfr1262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146974.1 | NP_667185.1 | 1 | 470 | ||
| GenBank mRNA | AB074429.1 | - | 1 | 470 | ||
| BC120560.1 | - | 1 | 531 | 40129483 | ||
| BC120562.1 | - | 1 | 531 | 40129485 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1262| Gene Symbol: Olfr1262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146974.1 | NP_667185.1 | 1 | 470 | ||
| GenBank mRNA | AB074429.1 | - | 1 | 470 | ||
| BC120560.1 | - | 1 | 531 | 40129483 | ||
| BC120562.1 | - | 1 | 531 | 40129485 | ||