siRNA Details
| Gene Symbol: Olfr639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147084.1 | NP_667295.1 | 1 | 142 | ||
| Gene Symbol: 4930516K23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015805.1 | - | 6 | 623 | ||

Olfr639
Genomic Map