siRNA Details
| Gene Symbol: Zar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174877.3 | NP_777366.1 | 1, 2 | 809 | ||
| GenBank mRNA | AY191415.1 | - | 1, 2 | 809 | ||
| BC099399.1 | - | 1, 2 | 803 | 30939000 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zar1| Gene Symbol: Zar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174877.3 | NP_777366.1 | 1, 2 | 809 | ||
| GenBank mRNA | AY191415.1 | - | 1, 2 | 809 | ||
| BC099399.1 | - | 1, 2 | 803 | 30939000 | ||