siRNA Details
| Gene Symbol: ZAK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016653.2 | NP_057737.2 | 2 | 281 | ||
| NM_133646.2 | NP_598407.1 | 2 | 281 | |||
| XM_005246640.2 | XP_005246697.1 | 2 | 327 | |||
| XM_017004323.1 | XP_016859812.1 | 2 | 327 | |||
| XM_017004324.1 | XP_016859813.1 | 2 | 351 | |||
| GenBank mRNA | AB030034.1 | - | 2 | 256 | ||
| AB041883.1 | - | 2 | 351 | |||
| AB049733.1 | - | 1 | 81 | |||
| AB049734.1 | - | 1 | 81 | |||
| AB208974.1 | - | 2 | 217 | |||
| AF238255.1 | - | 1 | 81 | |||
| AF251441.1 | - | 2 | 159 | |||
| AF325454.1 | - | 1 | 86 | |||
| AF465843.1 | - | 2 | 250 | |||
| AF480461.1 | - | 2 | 276 | |||
| AF480462.1 | - | 2 | 276 | |||
| AK056310.1 | - | 2 | 282 | |||
| AK291825.1 | - | 2 | 173 | |||
| BC001401.2 | - | 2 | 211 | 3051317 | ||
| DA993056.1 | - | 2 | 282 | |||

ZAK
Genomic Map