siRNA Details
Gene Symbol: KHK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000221.2 | NP_000212.1 | 4 | 906 | ||
NM_006488.2 | NP_006479.1 | 4 | 906 | |||
XM_005264294.3 | XP_005264351.1 | 5 | 1049 | |||
XM_005264296.3 | XP_005264353.1 | 4 | 892 | |||
XM_005264298.3 | XP_005264355.1 | 2 | 659 | |||
XM_006712008.3 | XP_006712071.1 | 5 | 1049 | |||
XM_006712009.3 | XP_006712072.1 | 5 | 1046 | |||
XM_006712010.3 | XP_006712073.1 | 4 | 914 | |||
XM_006712011.3 | XP_006712074.1 | 4 | 911 | |||
XM_006712012.3 | XP_006712075.1 | 4 | 911 | |||
XM_006712013.3 | XP_006712076.1 | 3 | 776 | |||
XM_006712014.3 | XP_006712077.1 | 2 | 659 | |||
XM_017004060.1 | XP_016859549.1 | 5 | 1046 | |||
XM_017004061.1 | XP_016859550.1 | 3 | 776 | |||
GenBank mRNA | AB209518.1 | - | 1 | 282 | ||
AK130033.1 | - | 4 | 423 | |||
AK223075.1 | - | 4 | 423 | |||
BC006233.2 | - | 4 | 844 | 3946893 | ||
BX648873.1 | - | 2 | 173 | |||
CR456801.1 | - | 4 | 389 | |||
X78677.1 | - | 2 | 185 | |||
X78678.1 | - | 4 | 397 |

More Information
Panther Classification:
Molecular Function -





Gene Ontology Categories:
