siRNA Details
| Gene Symbol: EPHB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004444.4 | NP_004435.3 | 11 | 2302 | ||
| XM_017011816.1 | XP_016867305.1 | 11 | 1959 | |||
| GenBank mRNA | AB209644.1 | - | 10 | 3012 | ||
| AK301224.1 | - | 6 | 609 | |||
| AY056048.1 | - | 11 | 1896 | |||
| BC004264.1 | - | 8 | 1387 | |||
| BC052804.1 | - | 11 | 1979 | 6584384 | ||
| U07695.1 | - | 11 | 1896 | |||

EPHB4
Genomic Map