siRNA Details
| Gene Symbol: ETNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018638.4 | NP_061108.2 | 2, 3 | 713 | ||
| XM_017019580.1 | XP_016875069.1 | 2, 3 | 710 | |||
| XM_017019581.1 | XP_016875070.1 | 2, 3 | 710 | |||
| GenBank mRNA | AF207600.2 | - | 2, 3 | 779 | ||
| AK074425.1 | - | 2, 3 | 658 | |||
| AL137750.1 | - | 2, 3 | 662 | |||
| BC066907.1 | - | 2, 3 | 713 | 5260109 | ||

ETNK1
Genomic Map