siRNA Details
| Gene Symbol: PANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138316.3 | NP_612189.2 | 2 | 456 | ||
| NM_148977.2 | NP_683878.1 | 2 | 1234 | |||
| NM_148978.2 | NP_683879.1 | 2 | 456 | |||
| XM_017016333.1 | XP_016871822.1 | 3 | 1202 | |||
| XM_017016334.1 | XP_016871823.1 | 3 | 1164 | |||
| XM_017016335.1 | XP_016871824.1 | 3 | 1992 | |||
| XM_017016336.1 | XP_016871825.1 | 2 | 381 | |||
| XM_017016337.1 | XP_016871826.1 | 3 | 1898 | |||
| GenBank mRNA | AF355198.1 | - | 2 | 440 | ||
| AK125428.1 | - | 3 | 813 | |||
| AK127777.1 | - | 1 | 155 | |||
| AY027661.1 | - | 2 | 1234 | |||
| AY027662.1 | - | 2 | 456 | |||
| AY027663.1 | - | 3 | 550 | |||

PANK1
Genomic Map