siRNA Details
| Gene Symbol: ICK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014920.3 | NP_055735.1 | 5 | 721 | ||
| NM_016513.4 | NP_057597.2 | 6 | 832 | |||
| XM_011514419.1 | XP_011512721.1 | 5 | 721 | |||
| XM_011514420.2 | XP_011512722.1 | 6 | 863 | |||
| XM_011514421.1 | XP_011512723.1 | 6 | 802 | |||
| XM_017010485.1 | XP_016865974.1 | 6 | 1095 | |||
| XM_017010486.1 | XP_016865975.1 | 7 | 974 | |||
| XM_017010487.1 | XP_016865976.1 | 5 | 640 | |||
| XM_017010488.1 | XP_016865977.1 | 6 | 1522 | |||
| XM_017010489.1 | XP_016865978.1 | 7 | 1037 | |||
| XM_017010490.1 | XP_016865979.1 | 6 | 907 | |||
| XM_017010491.1 | XP_016865980.1 | 6 | 868 | |||
| XM_017010492.1 | XP_016865981.1 | 5 | 912 | |||
| GenBank mRNA | AB023153.1 | - | 5 | 608 | ||
| AF152469.1 | - | 5 | 689 | |||
| AF225919.1 | - | 5 | 551 | |||
| AK074892.1 | - | 6 | 802 | |||
| BC035807.1 | - | 5 | 753 | |||
| BC136420.1 | - | 4 | 382 | 9020408 | ||
| BC136421.1 | - | 4 | 382 | 9020409 | ||
| BC152464.1 | - | 5 | 608 | 8862600 | ||

ICK
Genomic Map