siRNA Details
| Gene Symbol: RYK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005861.2 | NP_001005861.1 | 8 | 1059 | ||
| NM_002958.3 | NP_002949.2 | 8 | 1050 | |||
| XM_017007016.1 | XP_016862505.1 | 3 | 556 | |||
| GenBank mRNA | BC021700.2 | - | 3 | 325 | ||
| S59184.1 | - | 8 | 1083 | |||
| X96588.1 | - | 9 | 955 | |||

RYK
Genomic Map