siRNA Details
| Gene Symbol: SCYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317784.1 | NP_001304713.1 | 8 | 1367 | ||
| NM_017988.5 | NP_060458.3 | 8 | 1519 | |||
| XM_017019618.1 | XP_016875107.1 | 8 | 1367 | |||
| XM_017019619.1 | XP_016875108.1 | 8 | 1519 | |||
| XM_017019620.1 | XP_016875109.1 | 9 | 1506 | |||
| GenBank mRNA | AB037781.1 | - | 5 | 673 | ||
| AK000936.1 | - | 7 | 1125 | |||
| AK024274.1 | - | 4 | 528 | |||
| AK027551.1 | - | 4 | 585 | |||
| AK125091.1 | - | 8 | 1504 | |||
| AK292980.1 | - | 8 | 1293 | |||
| BC063798.1 | - | 8 | 1132 | 6473586 | ||

SCYL2
Genomic Map