siRNA Details
| Gene Symbol: INSRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014215.2 | NP_055030.1 | 3 | 1100 | ||
| GenBank mRNA | AK310760.1 | - | 3 | 1101 | ||
| J05046.1 | - | 2 | 716 | |||
| JQ991924.1 | - | 1 | 72 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

INSRR| Gene Symbol: INSRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014215.2 | NP_055030.1 | 3 | 1100 | ||
| GenBank mRNA | AK310760.1 | - | 3 | 1101 | ||
| J05046.1 | - | 2 | 716 | |||
| JQ991924.1 | - | 1 | 72 | |||