siRNA Details
| Gene Symbol: WNK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002838.3 | NP_001002838.1 | 2 | 567 | ||
| NM_020922.4 | NP_065973.2 | 2 | 567 | |||
| XM_006724593.3 | XP_006724656.1 | 2 | 505 | |||
| XM_011530802.2 | XP_011529104.1 | 2 | 505 | |||
| XM_017029741.1 | XP_016885230.1 | 2 | 506 | |||
| XM_017029742.1 | XP_016885231.1 | 2 | 583 | |||
| XM_017029743.1 | XP_016885232.1 | 2 | 498 | |||
| XM_017029744.1 | XP_016885233.1 | 2 | 400 | |||
| XM_017029745.1 | XP_016885234.1 | 2 | 505 | |||
| GenBank mRNA | AJ409088.2 | - | 1 | 128 | ||
| AY082340.1 | - | 1 | 128 | |||
| AY352048.1 | - | 2 | 567 | |||

WNK3
Genomic Map