siRNA Details
| Gene Symbol: NEK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178170.2 | NP_835464.1 | 9 | 1236 | ||
| XM_011524638.2 | XP_011522940.1 | 10 | 1479 | |||
| XM_011524640.2 | XP_011522942.1 | 10 | 1440 | |||
| XM_017024499.1 | XP_016879988.1 | 9 | 1285 | |||
| XM_017024500.1 | XP_016879989.1 | 9 | 1321 | |||
| XM_017024501.1 | XP_016879990.1 | 8, 9 | 1596 | |||
| GenBank mRNA | AK301429.1 | - | 8 | 1406 | ||
| AY242354.1 | - | 9 | 1236 | |||
| AY267371.1 | - | 9 | 1236 | |||
| BC112240.1 | - | 9 | 1270 | 8327708 | ||
| BC113705.1 | - | 9 | 1270 | 8322757 | ||

NEK8
Genomic Map