siRNA Details
| Gene Symbol: CDK19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300960.1 | NP_001287889.1 | 2 | 329 | ||
| NM_001300963.1 | NP_001287892.1 | 3 | 145 | |||
| NM_001300964.1 | NP_001287893.1 | 2 | 99 | |||
| NM_015076.4 | NP_055891.1 | 2 | 329 | |||
| XM_011535630.2 | XP_011533932.1 | 3 | 807 | |||
| XM_011535631.2 | XP_011533933.1 | 3 | 799 | |||
| XM_011535632.1 | XP_011533934.1 | 1 | 21 | |||
| XM_017010587.1 | XP_016866076.1 | 2 | 274 | |||
| GenBank mRNA | AB028951.2 | - | 2 | 146 | ||
| AK292360.1 | - | 2 | 237 | |||
| AY028424.1 | - | 2 | 329 | |||
| BC024247.1 | - | 3 | 150 | |||
| BC037289.1 | - | 2 | 224 | 5266531 | ||
| DB030002.1 | - | 2 | 99 | |||
| JX028238.1 | - | 2 | 149 | |||

CDK19
Genomic Map