siRNA Details
| Gene Symbol: CIT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206999.1 | NP_001193928.1 | 46 | 5860 | ||
| NM_007174.2 | NP_009105.1 | 45 | 5734 | |||
| XM_006719206.2 | XP_006719269.1 | 46 | 5842 | |||
| XM_011537783.1 | XP_011536085.1 | 47 | 5932 | |||
| XM_011537784.1 | XP_011536086.1 | 47 | 5932 | |||
| XM_011537785.1 | XP_011536087.1 | 47 | 5887 | |||
| XM_011537787.1 | XP_011536089.1 | 45 | 5716 | |||
| XM_011537788.1 | XP_011536090.1 | 45 | 5716 | |||
| XM_011537789.1 | XP_011536091.1 | 39 | 5067 | |||
| XM_011537790.2 | XP_011536092.1 | 38 | 4635 | |||
| XM_011537791.2 | XP_011536093.1 | 38 | 5620 | |||
| XM_011537792.2 | XP_011536094.1 | 37 | 5675 | |||
| XM_017018735.1 | XP_016874224.1 | 46 | 5887 | |||
| XM_017018736.1 | XP_016874225.1 | 47 | 5887 | |||
| XM_017018737.1 | XP_016874226.1 | 46 | 5842 | |||
| XM_017018738.1 | XP_016874227.1 | 36 | 5176 | |||
| XM_017018739.1 | XP_016874228.1 | 36 | 5951 | |||
| GenBank mRNA | AB023166.2 | - | 37 | 5675 | ||
| AY257469.1 | - | 45 | 5731 | |||
| AY681966.1 | - | 46 | 5860 | |||

CIT
Genomic Map