siRNA Details
| Gene Symbol: ANKK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178510.1 | NP_848605.1 | 4, 5 | 784 | ||
| XM_011542737.2 | XP_011541039.1 | 4, 5 | 2561 | |||
| GenBank mRNA | AF487542.1 | - | 4, 5 | 690 | ||
| AJ541797.2 | - | 4, 5 | 784 | |||

ANKK1
Genomic Map