siRNA Details
| Gene Symbol: CAPNS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003962.2 | NP_001003962.1 | 7 | 738 | ||
| NM_001302632.1 | NP_001289561.1 | 7 | 645 | |||
| NM_001302633.1 | NP_001289562.1 | 6 | 430 | |||
| NM_001749.3 | NP_001740.1 | 7 | 741 | |||
| XM_005259295.1 | XP_005259352.1 | 7 | 541 | |||
| XM_005259296.1 | XP_005259353.1 | 7 | 544 | |||
| XM_011527359.1 | XP_011525661.1 | 6 | 371 | |||
| GenBank mRNA | AK289606.1 | - | 7 | 680 | ||
| AY007141.1 | - | 7 | 538 | |||
| BC000592.2 | - | 7 | 559 | 3347146 | ||
| BC007779.2 | - | 7 | 569 | 4301394 | ||
| BC011903.2 | - | 7 | 559 | 4120579 | ||
| BC017308.2 | - | 7 | 538 | 5089022 | ||
| BC018931.2 | - | 7 | 629 | 4303926 | ||
| BC021933.1 | - | 7 | 533 | 4872635 | ||
| BC023643.2 | - | 7 | 531 | 5017512 | ||
| BC064998.1 | - | 7 | 646 | 5785274 | ||
| BQ891655.1 | - | 4 | 202 | |||
| BT009775.1 | - | 6 | 494 | |||
| CN349928.1 | - | 6 | 379 | |||
| X04106.1 | - | 7 | 652 | |||

CAPNS1
Genomic Map