siRNA Details
| Gene Symbol: PRSS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003619.3 | NP_003610.2 | 12 | 2167 | ||
| GenBank mRNA | AF077298.1 | - | 3 | 278 | ||
| AJ001531.1 | - | 12 | 2160 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRSS12| Gene Symbol: PRSS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003619.3 | NP_003610.2 | 12 | 2167 | ||
| GenBank mRNA | AF077298.1 | - | 3 | 278 | ||
| AJ001531.1 | - | 12 | 2160 | |||