siRNA Details
| Gene Symbol: CPVL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019029.2 | NP_061902.2 | 13 | 1554 | ||
| NM_031311.3 | NP_112601.3 | 13 | 1466 | |||
| XM_011515437.1 | XP_011513739.1 | 13 | 1633 | |||
| XM_017012366.1 | XP_016867855.1 | 11 | 1193 | |||
| GenBank mRNA | AF106704.2 | - | 13 | 1386 | ||
| AF217508.1 | - | 12 | 1354 | |||
| AF282617.1 | - | 13 | 1416 | |||
| AK075433.1 | - | 13 | 1467 | |||
| AK124472.1 | - | 17 | 1994 | |||
| AK225545.1 | - | 13 | 1417 | |||
| AY358549.1 | - | 13 | 1443 | |||
| BC016838.2 | - | 13 | 1514 | 3888647 | ||

CPVL
Genomic Map