siRNA Details
| Gene Symbol: TMPRSS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077263.2 | NP_001070731.1 | 13 | 1837 | ||
| NM_001206789.1 | NP_001193718.1 | 12 | 1732 | |||
| GenBank mRNA | AB048797.1 | - | 12 | 1688 | ||
| AK300283.1 | - | 13 | 1749 | |||
| BC018715.1 | - | 14 | 1572 | |||
| BC114928.1 | - | 13 | 1756 | 40028749 | ||

TMPRSS13
Genomic Map