siRNA Details
Gene Symbol: DPP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320875.1 | NP_001307804.1 | 5, 6 | 921 | ||
NM_001320876.1 | NP_001307805.1 | 4, 5 | 890 | |||
NM_017743.5 | NP_060213.2 | 4, 5 | 785 | |||
NM_130434.4 | NP_569118.1 | 4, 5 | 785 | |||
NM_197960.3 | NP_932064.1 | 5, 6 | 826 | |||
NM_197961.3 | NP_932065.1 | 5, 6 | 921 | |||
NR_135485.1 | 4, 5 | 785 | ||||
NR_135486.1 | 5, 6 | 826 | ||||
XM_011521730.1 | XP_011520032.1 | 4, 5 | 2174 | |||
XM_011521732.2 | XP_011520034.1 | 4, 5 | 2174 | |||
XM_011521734.1 | XP_011520036.1 | 4, 5 | 2174 | |||
XM_017022373.1 | XP_016877862.1 | 4, 5 | 2174 | |||
XM_017022374.1 | XP_016877863.1 | 4, 5 | 2174 | |||
XM_017022375.1 | XP_016877864.1 | 4, 5 | 766 | |||
XM_017022376.1 | XP_016877865.1 | 4, 5 | 766 | |||
XM_017022378.1 | XP_016877867.1 | 4, 5 | 2174 | |||
GenBank mRNA | AF221634.1 | - | 4, 5 | 759 | ||
AK000290.1 | - | 4, 5 | 708 | |||
AK291057.1 | - | 5, 6 | 896 | |||
AK296915.1 | - | 4, 5 | 861 | |||
AK302638.1 | - | 5, 6 | 762 | |||
AY172659.1 | - | 3, 4 | 553 | |||
AY354202.1 | - | 5, 6 | 754 | |||
BC030688.2 | - | 5, 6 | 912 | 4822550 |
