siRNA Details
| Gene Symbol: AEBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129.4 | NP_001120.3 | 11 | 1588 | ||
| XM_011515162.1 | XP_011513464.1 | 10 | 1551 | |||
| GenBank mRNA | AB209744.1 | - | 11 | 1542 | ||
| AF053944.1 | - | 11 | 1422 | |||
| BC038588.1 | - | 11 | 1588 | 5764769 | ||
| D86479.1 | - | 5 | 344 | |||

AEBP1
Genomic Map