siRNA Details
| Gene Symbol: ADAM20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003814.4 | NP_003805.3 | 2 | 2395 | ||
| XM_005268151.3 | XP_005268208.1 | 2 | 11291 | |||
| GenBank mRNA | AF029899.1 | - | 1 | 2028 | ||
| BC025378.1 | - | 2 | 2398 | 4827383 | ||

ADAM20
Genomic Map