siRNA Details
| Gene Symbol: AFG3L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006796.2 | NP_006787.2 | 15 | 2006 | ||
| GenBank mRNA | AK129504.1 | - | 8 | 832 | ||
| BC024282.1 | - | 16 | 1859 | |||
| BC065016.1 | - | 15 | 1928 | 6169804 | ||
| Y18314.1 | - | 15 | 1925 | |||

AFG3L2
Genomic Map