siRNA Details
| Gene Symbol: PAPPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020318.2 | NP_064714.2 | 2, 3 | 2091 | ||
| NM_021936.2 | NP_068755.2 | 2, 3 | 2091 | |||
| XM_005245422.3 | XP_005245479.1 | 2, 3 | 2138 | |||
| XM_011509857.1 | XP_011508159.1 | 3, 4 | 2008 | |||
| XM_011509858.2 | XP_011508160.1 | 2, 3 | 2351 | |||
| XM_017002023.1 | XP_016857512.1 | 3, 4 | 2189 | |||
| XM_017002024.1 | XP_016857513.1 | 3, 4 | 2236 | |||
| XM_017002025.1 | XP_016857514.1 | 2, 3 | 2342 | |||
| GenBank mRNA | AF311940.1 | - | 1, 2 | 927 | ||
| AF342989.1 | - | 2, 3 | 2091 | |||
| AF342990.1 | - | 2, 3 | 2091 | |||
| AJ278348.1 | - | 1, 2 | 890 | |||
| BC117193.1 | - | 1, 2 | 1699 | |||
| BC117195.1 | - | 1, 2 | 1700 | |||
| BC152552.1 | - | 2, 3 | 2079 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001085376, AK084006.

PAPPA2
Genomic Map