siRNA Details
| Gene Symbol: NLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020726.4 | NP_065777.1 | 2 | 429 | ||
| XM_005248559.2 | XP_005248616.1 | 2 | 420 | |||
| XM_006714661.2 | XP_006714724.1 | 3 | 484 | |||
| XM_017009673.1 | XP_016865162.1 | 3 | 484 | |||
| GenBank mRNA | AB033052.2 | - | 2 | 251 | ||
| AJ300837.1 | - | 2 | 376 | |||
| AK096416.1 | - | 2 | 374 | |||
| AK292803.1 | - | 2 | 370 | |||
| BC001644.2 | - | 2 | 367 | 2958628 | ||
| BC004985.1 | - | 2 | 367 | 2958628 | ||
| CB992921.1 | - | 2 | 456 | |||

NLN
Genomic Map