siRNA Details
| Gene Symbol: AFG3L1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003226.1 | 11 | 2652 | |||
| NR_003227.1 | 10 | 2413 | ||||
| GenBank mRNA | AK056488.1 | - | 12 | 2258 | ||
| AK090856.1 | - | 2 | 1863 | |||
| AK091293.1 | - | 11 | 2615 | |||
| AK097044.1 | - | 10 | 2400 | |||
| AK126911.1 | - | 9 | 2443 | |||
| AK128352.1 | - | 4 | 2252 | |||
| AK128804.1 | - | 5 | 1779 | |||
| AL832984.1 | - | 5 | 1876 | |||
| BC021245.2 | - | 9 | 2225 | |||

AFG3L1P
Genomic Map