siRNA Details
| Gene Symbol: ADAM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318933.1 | NP_001305862.1 | 27 | 2902 | ||
| NM_002390.5 | NP_002381.2 | 27 | 2906 | |||
| XM_005257373.3 | XP_005257430.1 | 26 | 3547 | |||
| XM_017024647.1 | XP_016880136.1 | 25 | 3319 | |||
| GenBank mRNA | AB009675.1 | - | 27 | 2885 | ||
| BM557705.1 | - | 1 | 401 | |||
| D17390.1 | - | 24 | 2625 | |||

ADAM11
Genomic Map