siRNA Details
| Gene Symbol: PRSS36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258290.1 | NP_001245219.1 | 15 | 2805 | ||
| NM_001258291.1 | NP_001245220.1 | 14 | 2511 | |||
| NM_173502.4 | NP_775773.2 | 15 | 2820 | |||
| XM_011545758.2 | XP_011544060.1 | 14 | 3038 | |||
| XM_017022971.1 | XP_016878460.1 | 12 | 2279 | |||
| GenBank mRNA | AJ627034.1 | - | 15 | 2792 | ||
| AK075142.1 | - | 15 | 2793 | |||
| AK290310.1 | - | 15 | 2820 | |||
| AK297997.1 | - | 13 | 2761 | |||

PRSS36
Genomic Map