siRNA Details
| Gene Symbol: CLCN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256944.1 | NP_001243873.1 | 3, 4 | 615 | ||
| NM_001830.3 | NP_001821.2 | 5, 6 | 870 | |||
| GenBank mRNA | AB019432.1 | - | 4, 5 | 715 | ||
| AF170492.1 | - | 3, 4 | 440 | |||
| AK225287.1 | - | 5, 6 | 831 | |||
| AK289564.1 | - | 5, 6 | 817 | |||
| AK299611.1 | - | 2, 3 | 248 | |||
| BC036068.1 | - | 4, 5 | 490 | 5311366 | ||
| BC130278.1 | - | 4, 5 | 729 | 40146309 | ||
| X77197.1 | - | 5, 6 | 823 | |||

CLCN4
Genomic Map