siRNA Details
| Gene Symbol: GLRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001118885.1 | NP_001112357.1 | 10 | 1590 | ||
| NM_001118886.1 | NP_001112358.1 | 9 | 2018 | |||
| NM_001171942.1 | NP_001165413.1 | 11 | 1757 | |||
| NM_002063.3 | NP_002054.1 | 9 | 2018 | |||
| XM_006724487.3 | XP_006724550.1 | 10 | 1401 | |||
| XM_011545495.2 | XP_011543797.1 | 11 | 2075 | |||
| XM_011545496.2 | XP_011543798.1 | 11 | 1785 | |||
| XM_017029427.1 | XP_016884916.1 | 10 | 1562 | |||
| XM_017029428.1 | XP_016884917.1 | 10 | 1436 | |||
| XM_017029429.1 | XP_016884918.1 | 11 | 1981 | |||
| GenBank mRNA | AK289561.1 | - | 9 | 1545 | ||
| AK297268.1 | - | 9 | 1509 | |||
| AK297304.1 | - | 11 | 1694 | |||
| AK312591.1 | - | 10 | 1590 | |||
| AY437083.1 | - | 9 | 1258 | |||
| AY437084.1 | - | 9 | 1258 | |||
| BC032864.2 | - | 9 | 1788 | 5263356 | ||
| CR450343.1 | - | 9 | 1258 | |||
| X52008.1 | - | 9 | 1645 | |||

GLRA2
Genomic Map