siRNA Details
Gene Symbol: KCNJ10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002241.4 | NP_002232.2 | 2 | 2223 | ||
GenBank mRNA | BC034036.1 | - | 2 | 2125 | ||
BC131627.1 | - | 2 | 2119 | 40118533 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KCNJ10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002241.4 | NP_002232.2 | 2 | 2223 | ||
GenBank mRNA | BC034036.1 | - | 2 | 2125 | ||
BC131627.1 | - | 2 | 2119 | 40118533 |