siRNA Details
| Gene Symbol: KCNK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003740.3 | NP_003731.1 | 5 | 3462 | ||
| XM_005249456.1 | XP_005249513.1 | 5 | 3186 | |||
| XM_006715235.2 | XP_006715298.1 | 3 | 2725 | |||
| GenBank mRNA | AF084830.1 | - | 6 | 3158 | ||
| AK001897.1 | - | 1 | 2059 | |||
| BC060793.1 | - | 5 | 3462 | 30348251 | ||

KCNK5
Genomic Map