siRNA Details
| Gene Symbol: KCNK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003740.3 | NP_003731.1 | 5 | 3639 | ||
| XM_005249456.1 | XP_005249513.1 | 5 | 3363 | |||
| XM_006715235.2 | XP_006715298.1 | 3 | 2902 | |||
| GenBank mRNA | AF084830.1 | - | 6 | 3335 | ||
| AK001897.1 | - | 1 | 2236 | |||

KCNK5
Genomic Map