siRNA Details
| Gene Symbol: BCL7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197244.1 | NP_001184173.1 | 5 | 983 | ||
| NM_001301061.1 | NP_001287990.1 | 7 | 1276 | |||
| NM_001707.3 | NP_001698.2 | 6 | 1154 | |||
| NR_036682.1 | 6 | 1137 | ||||
| GenBank mRNA | AJ223979.1 | - | 6 | 553 | ||
| BC000956.2 | - | 6 | 1113 | 3450946 | ||
| BC001967.2 | - | 6 | 1020 | 3461829 | ||
| BC009548.1 | - | 6 | 1064 | 3897805 | ||
| X89985.1 | - | 6 | 1031 | |||

BCL7B
Genomic Map