siRNA Details
| Gene Symbol: KCNC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004977.2 | NP_004968.2 | 5 | 2659 | ||
| GenBank mRNA | AF055989.1 | - | 5 | 2659 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNC3| Gene Symbol: KCNC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004977.2 | NP_004968.2 | 5 | 2659 | ||
| GenBank mRNA | AF055989.1 | - | 5 | 2659 | ||