siRNA Details
| Gene Symbol: CFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001710.5 | NP_001701.2 | 18 | 2563 | ||
| GenBank mRNA | AK130533.1 | - | 18 | 2492 | ||
| AK223400.1 | - | 18 | 2411 | |||
| BC004143.2 | - | 18 | 2420 | 2959705 | ||
| BC007990.2 | - | 18 | 2420 | 2959706 | ||
| K01566.1 | - | 17 | 2024 | |||
| L15702.1 | - | 18 | 2325 | |||
| S67310.1 | - | 18 | 2396 | |||
| X72875.1 | - | 18 | 2285 | |||

CFB
Genomic Map