siRNA Details
| Gene Symbol: KCND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004979.5 | NP_004970.3 | 4 | 2987 | ||
| XM_011543910.2 | XP_011542212.1 | 5 | 1895 | |||
| XM_017029508.1 | XP_016884997.1 | 5 | 1726 | |||
| XM_017029509.1 | XP_016884998.1 | 5 | 1723 | |||
| GenBank mRNA | AB021865.1 | - | 4 | 2712 | ||
| AF166003.1 | - | 4 | 1464 | |||
| AJ005898.1 | - | 4 | 1414 | |||
| AK094431.1 | - | 3 | 1573 | |||
| AK315092.1 | - | 4 | 1898 | |||
| BC045659.1 | - | 4 | 2985 | 5271042 | ||

KCND1
Genomic Map